missing translation for 'onlineSavingsMsg'
Learn More

Napropamid, TRC

Product Code. 30465774
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30465774 10 g
30474839 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30465774 Supplier Toronto Research Chemicals Supplier No. TRCN37870010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15299-99-7
Molecular Formula C17 H21 N O2
Molecular Weight (g/mol) 271.35
Synonym Propionamide, N,N-diethyl-2-(1-naphthyloxy)- (8CI), N,N-Diethyl-2-(1-naphthalenyloxy)propanamide, (±)-Devrinol, 2-(α-Naphthoxy)-N,N-diethylpropionamide, Devrinol, N,N-Diethyl-2-(1-naphthalenyloxy)propionamide, N,N-Diethyl-2-(1-naphthyloxy)propanamide, Napropamid, Napropamide, R 7465, Racemic devrinol
IUPAC Name N,N-diethyl-2-naphthalen-1-yloxypropanamide
SMILES CCN(CC)C(=O)C(C)Oc1cccc2ccccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H21NO2/c1-4-18(5-2)17(19)13(3)20-16-12-8-10-14-9-6-7-11-15(14)16/h6-13H,4-5H2,1-3H3
Formula Weight 271.1572
Chemical Name or Material Napropamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.