missing translation for 'onlineSavingsMsg'
Learn More

Methyl quinoline-4-carboxylate (~90%), TRC

Product Code. 30466039
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466039 50 mg 50mg
30483584 5 mg 5mg
30479350 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30466039 Supplier Toronto Research Chemicals Supplier No. TRCM34235350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21233-61-4
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.19
Synonym Methyl quinoline-4-carboxylate
IUPAC Name methyl quinoline-4-carboxylate
SMILES COC(=O)c1ccnc2ccccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H9NO2/c1-14-11(13)9-6-7-12-10-5-3-2-4-8(9)10/h2-7H,1H3
Formula Weight 187.0633
Chemical Name or Material Methyl quinoline-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.