missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenedianiline, TRC

Product Code. 30468237
Change view
Click to view available options
Quantity:
1 g
5 g
250 g
Unit Size:
1g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468237 250 g 250g
30465482 5 g 5g
30469843 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468237 Supplier Toronto Research Chemicals Supplier No. TRCM304180250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-77-9
Molecular Formula C13 H14 N2
Molecular Weight (g/mol) 198.26
Synonym Aniline, 4,4'-methylenedi- (8CI), Aniline, p,p'-methylenebis- (3CI), 4,4'-Methylenebis[benzenamine], 4,4-Diaminodiphenylmethane, 4,4'-Diaminobiphenylmethane, 4,4'-Diaminodiphenylenemethane, 4,4'-Diaminodiphenylmethane, 4,4'-Diaminophenylmethane, 4,4'-Diphenylmethanediamine, 4,4'-Methylenebis(aniline), 4,4'-Methylenebisbenzeneamine, 4,4'-Methylenedianiline, 4,4'-Methylenedibenzenamine, 4,4'-Methylenediphenylamine, 4-(4-Aminobenzyl)aniline, 4-(4-Aminobenzyl)benzenamine, 4-(p-Aminobenzyl)aniline, Acmex H 84BM, Amicure 101, Ancamine 1692, Ancamine TL, Aradur HT 972GB, Araldite HT 972, Araldite HY 972, Araldite Hardener 972, Araldite Hardener HT 972, Araldite R 972, Bis(4-aminophenyl)methane, Bis(aminophenyl)methane, Bis(p-aminophenyl)methane, DAM, DDM, DEH 50, Di(4-aminophenyl)methane, Diaminodiphenylmethane, Dianilinemethane, Dianilinomethane, Epicure DDM, HT 972, Kayahard PT-AA, MDA, MDA 100, MDA 2200, Methylenedianiline, NSC 4709, Sumicure M, Tonox, Tonox MDA, Wanamine MDA, XU-HY 205, p,p'-Diaminodiphenylmethane, p,p'-Methylenedianiline
IUPAC Name 4-[(4-aminophenyl)methyl]aniline
SMILES Nc1ccc(Cc2ccc(N)cc2)cc1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C13H14N2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8H,9,14-15H2
Formula Weight 198.1157
Chemical Name or Material Bis-(4-aminophenyl)methane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.