missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methylpyridine-3-boronic Acid Hydrochloride Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Methylpyridine-3-boronic Acid Hydrochloride Hydrate |
| Analyte or Component Names | 6-Methylpyridine-3-boronic Acid Hydrochloride Hydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6H8BNO2. HCl . xH 2O |
| Synonym | B-(2-Methylpyridin-5-yl)boronic Acid Hydrochloride Hydrate, (2-Methylpyridin-5-yl)boronic Acid Hydrochloride Hydrate, (6-Methyl-3-pyridinyl)boronic Acid Hydrochloride Hydrate, 2-Methyl-5-pyridinylboronic Acid Hydrochloride Hydrate, 6-Methyl-3-pyridineboronic Acid Hydrochloride Hydrate |
| InChI Formula | InChI=1S/C6H8BNO2.ClH.H2O/c1-5-2-3-6(4-8-5)7(9)10;;/h2-4,9-10H,1H3;1H;1H2 |
| Solution Type | Neat |
| SMILES | OB(C1=CC=C(N=C1)C)O.Cl.O |
| IUPAC Name | (6-methylpyridin-3-yl)boronic acid hydrochloride hydrate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?