missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-naphthalenediol, TRC

Product Code. 30481337
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481337 100 mg 100mg
30465684 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481337 Supplier Toronto Research Chemicals Supplier No. TRCM323135100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-85-6
Molecular Formula C11 H10 O2
Molecular Weight (g/mol) 174.2
Synonym 1,4-Dihydroxy-2-methylnaphthalene, 2-Methyl-1,4-dihydroxynaphthalene, 2-Methyl-1,4-naphthohydroquinone, 2-Methyl-1,4-naphthoquinol, 2-Methylhydronaphthoquinone, 2-Methylnaphthalene-1,4-diol, 4-Hydroxy-3-methyl-α-naphthol, Dihydrovitamin K3, Menadiol, Reduced Menadione, Reduced Vitamin K3, Vitamin K3H2
IUPAC Name 2-methylnaphthalene-1,4-diol
SMILES Cc1cc(O)c2ccccc2c1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H10O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6,12-13H,1H3
Formula Weight 174.0681
Chemical Name or Material 2-Methyl-1,4-naphthalenediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.