missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3,5-Tri-O-acetyl-D-ribofuranoside, TRC

Product Code. 30481618
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481618 5 g 5g
30482474 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481618 Supplier Toronto Research Chemicals Supplier No. TRCM3307755G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52554-28-6
Molecular Formula C12H18O8
Molecular Weight (g/mol) 290.27
IUPAC Name [(2R,3R,4R)-3,4-diacetyloxy-5-methoxyoxolan-2-yl]methyl acetate
SMILES COC1O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H18O8/c1-6(13)17-5-9-10(18-7(2)14)11(19-8(3)15)12(16-4)20-9/h9-12H,5H2,1-4H3/t9-,10-,11-,12?/m1/s1
Formula Weight 290.1002
Chemical Name or Material Methyl 2,3,5-Tri-O-acetyl-D-ribofuranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.