missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve, ISO Certified, Pore Sizes: 10mm and Larger

Brand:  RETSCH 60.158.011200

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10014951

  • 251.00 € / Each

Estimated Shipment: 22-06-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
305mm
1Each
Test Sieve
Stainless Steel
Certificate indicating Compliance
Stainless Steel
For separation, fractioning, particle size determination
11.20mm
ISO 3310/1
Shaker AS 300
40mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers