Learn More
Skytec™ 3530 - P3 Disposable Pre-Formed Cup Shape Respirator
Product Code. 30242992 Shop All Skytec Products
Change view
Click to view available options
Quantity:
5/Pk.
Unit Size:
Pack of 5
Product Code. Quantity Unit Size
30242992 5/Pk. Pack of 5
1 options
Product Code. 30242992 Supplier Skytec™ Supplier No. ASR35300AZ
  • ONLINE EXCLUSIVE PRICE
    46.49 €
    30.97 € / Pack of 5
    Save 15.52 €
    33% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The 3500 SERIES disposable respirators offer an outstanding level of protection demonstrated when conducting Face Fit Testing. Having electrostatic filter media allows low breathing resistance and a lightweight outer shell maintains excellent shape.
  • Lightweight, cup shaped respirator
  • Four point headstrap adjustment
  • Preformed nosebridge removes the need for regular adjustment
  • Electrostatic filter media
  • Soft touch elastic
  • PVC Foam Inner Face Seal
  • Conforms to EN 149:2001+A1:2009 FFP3 NR
  • Low Profile Exhalation Valve helps reduce heat and moisture build up
Specifications
Aerosol Type R - Somewhat Resistant to Oil
Assigned Protection Factor (APF) 20
Certifications/Compliance CE III , UKCA , UK Medical Device Directive: 93/42/EEC and 2007/47/EC , EU Medical Device Regulation: 2017/745 , EN 149:2000 + A1:2009 FFP3 NR
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve No
Filtration Layer Material Melt Blown Polypropylene, Polyethylene
Material Headstrap Latex-Free Synthetic Rubber
Nose Piece Material PVC
Quantity 5/Pk.
Sterility Non-sterile