missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AX-024 hydrochloride, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Description
AX-024 hydrochloride is an orally available, first-in-class inhibitor of the TCR-Nck interaction that selectively inhibits TCR-triggered T cell activation with an IC50 ∼1 nM. AX-024 hydrochloride modulates cell signaling by targeting SH3 domains. AX-024 hydrochloride has low-acute toxicity and high potency and selectivity, and strongly inhibit the production of IL-6, TNF-α, IFN-γ, IL-10 and IL-17A.
Specifications
Specifications
| Chemical Name or Material | AX-024 hydrochloride |
| CAS | 1704801-24-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H23ClFNO2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 33 mg/mL (87.80 mM; Need ultrasonic and warming) ∣H2O : 16.67 mg/mL (44.35 mM; Need ultrasonic) |
| SMILES | COC1=CC=C(OC2)C(C(C3=CC=C(F)C=C3)=C2CN4CCCC4)=C1.[H]Cl |
| Molecular Weight (g/mol) | 375.86 |
| Formula Weight | 375.86 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction