Learn More
Corning™ 384-Well Black/Clear Round Bottom Ultra-Low Attachment Spheroid Microplate with Generic Bar codes, With Lid, Sterile
GREENER_CHOICE
Product Code. 16637962 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Product Code. Color
16637962 Black, Clear Bottom
1 options
Product Code. 16637962 Supplier Corning™ Supplier No. 3830BC
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
Ultra-Low Attachment microplates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation and cellular activation
  • Surface is noncytotoxic, biologically inert and nondegradable
  • Sterilized by gamma irradiation
  • Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk
  • Opaque walls to prevent well-to-well crosstalk
  • Can be used for both top and bottom reading instruments
  • Bulk packed 10 per bag
  • Novel well geometry aids formation of spheroids in center of well
  • Optically clear round bottom with black opaque microplate body
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Lid With Lid
Color Black, Clear Bottom
Volume (Metric) Well 90 μL
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Round
Quantity 10/Pk., 50/Cs.