Learn More
Corning™ 384 Well Flat Clear Bottom Polystyrene NBS™ Microplates, low flange, no lid, nonsterile
Product Code. 15575283 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 100
Product Code. Color Unit Size
15575283 Black Pack of 100
1 options
Product Code. 15575283 Supplier Corning™ Supplier No. 3766
  • ONLINE EXCLUSIVE PRICE
    1730.00 €
    1612.30 € / Pack of 100
    Save 117.70 €
    6% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise. Corning™ 384 Well Flat Clear Bottom Polystyrene NBS™ Microplates feature black wall plates that have low background fluorescence, minimal light scatter, and reduced crosstalk.
  • Black wall plates have low background fluorescence, minimal light scatter, and reduced crosstalk
  • Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimized molecular interactions
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
  • Suited for fluorescent assays using either top or bottom detection plate readers
  • Recommended working volume of up to 20 to 80μL
  • Conforms to standard microplate footprint and dimensions
TRUSTED_SUSTAINABILITY
Specifications
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Non-sterile
Surface Treatment Non-binding
Well Shape Square, Flat Bottom (F-well)
No. per Pack 25