missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96-Well Plate Plastic Non-Coated, Mid Height, Round Well
Shop All Thermo Scientific ProductsDescription
Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 96-Well Microplates. Made of high-quality, GC-tested polypropylene, these microplates are ideal for:
- Pharmaceutical and industrial QA/QC
- High-throughput screening (HTS)
- Sample collection and storage
- Combinatorial chemistry
- Liquid phase assays
Chemically and Thermally Resistant Polypropylene
- Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
- Will tolerate temperatures from −80° C to +121°C
- GC-tested to ensure low extractables
Broad Instrument Compatibility
- SBS and ANSI standard footprint design
C17
- Shallow well and deep well formats
- U- and V- bottom wells ensure optimal sample recovery
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Product Line | Webseal |
| Material | Plastic |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Non-coated |
| Well Shape | Round Bottom (U-well) |
| No. per Pack | 96 |
| Quantity | 96/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privatlivspolitik.
Ser du en mulighed for forbedring?