missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
GREENER_CHOICE

Product Code. 17880597 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Unit Size:
Pack of 5
Pack of 8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17880597 200 L Pack of 5
17840597 50 L Pack of 8
17860597 100 L Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17880597 Supplier Thermo Scientific™ Supplier No. SH30988.03
  • ONLINE EXCLUSIVE PRICE
    1995.00 €
    1682.48 € / Pack of 5
    Save 312.52 €
    15% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific 3D Tank Liners are composed of Aegis 5-14 film with dimensions optimized for Thermo Scientific drums and commonly-used, industry-standard cylindrical tanks. The tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 200 L
Quantity 200 L
Tubing Style C-Flex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.