Learn More
3M™ Versaflo™ Supplied Air Cooling Valve Assembly, V-100E
Product Code. 31605233 Shop All 3M Products
Change view
Click to view available options
Product Type:
Cooling Valve Assembly
Unit Size:
Each
Product Code. Product Type Unit Size
31605233 Cooling Valve Assembly Each
1 options
Product Code. 31605233 Supplier 3M™ Supplier No. 7100341417
  • ONLINE EXCLUSIVE PRICE
    1180.00 €
    752.31 € / Pack of 1
    Save 427.69 €
    36% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
V-Series supplied air regulators provide comfortable, cost effective respiratory protection for all-day use.
  • Supplied air respirators provide enhanced features compared to traditional respiratory protection.
  • Offer options to cool or heat incoming air with specific valves for added comfort.
  • Require a clean air source, such as plant air systems or bottled air, for operation.
  • Deliver clean, breathable air to a variety of compatible hoods and helmets.
  • Designed for low maintenance and reliable performance in demanding environments.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Product Type Cooling Valve Assembly
Size One Size