Learn More
3M™ Versaflo™ Supplied Air Heating Valve Assembly, V-200E
Product Code. 31605234 Shop All 3M Products
Change view
Click to view available options
Product Type:
Heating Valve Assembly
Unit Size:
Each
Product Code. Product Type Unit Size
31605234 Heating Valve Assembly Each
1 options
Product Code. 31605234 Supplier 3M™ Supplier No. 7100340967
  • ONLINE EXCLUSIVE PRICE
    1550.00 €
    989.18 € / Pack of 1
    Save 560.82 €
    36% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Supplied air heating valve assembly provides heated airflow for comfort in cold environments while ensuring reliable respiratory protection.
  • Provides a comfortable respiratory solution for workers in cold and challenging environments.
  • Heats compressed air by up to 28°C to enhance user comfort.
  • Compatible with a wide range of 3M headtops, including hoods and helmets.
  • Allows easy configuration based on environment, contaminants, and personal preferences.
  • Features adjustable airflow control to customize heating levels.
  • Suitable for use in manufacturing, construction, and other demanding industries.
  • Helps improve worker safety and overall comfort during extended use.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Product Type Heating Valve Assembly
Size One Size