Learn More
3M™ Versaflo™ Supplied Air Regulator Assembly, V-500E
Product Code. 31605235 Shop All 3M Products
Change view
Click to view available options
Product Type:
Adjustable Regulator
Unit Size:
Each
Product Code. Product Type Unit Size
31605235 Adjustable Regulator Each
1 options
Product Code. 31605235 Supplier 3M™ Supplier No. 7100342003
  • ONLINE EXCLUSIVE PRICE
    820.00 €
    524.67 € / Pack of 1
    Save 295.33 €
    36% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
V-500E Series supplied air regulator assembly provides reliable respiratory protection with adjustable airflow and continuous compressed air for comfort and durability in tough environments.
  • 3M™ Versaflo™ Supplied Air Respirator System V-500E Series provides reliable respiratory protection and comfort for workers across various industrial settings.
  • Delivers a continuous flow of air from a compressed air source, ensuring a steady and consistent air supply.
  • Compatible with a wide range of 3M headtops, including hoods and helmets, for flexible and versatile use.
  • Features adjustable airflow control, allowing users to customize airflow according to their specific needs and comfort preferences.
  • Offers effective protection against airborne contaminants for enhanced workplace safety.
  • Suitable for industries such as manufacturing, construction, and chemical processing.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Product Type Adjustable Regulator