Learn More
3M™ Versaflo™ Supplied Air Regulator Heavy Industry Kit, V-500E-HIK
Product Code. 31605229 Shop All 3M Products
Change view
Click to view available options
Product Type:
Supplied Air Regulator
Unit Size:
Each
Product Code. Product Type Unit Size
31605229 Supplied Air Regulator Each
1 options
Product Code. 31605229 Supplier 3M™ Supplier No. 7100341359
  • ONLINE EXCLUSIVE PRICE
    2005.00 €
    1282.58 € / Pack of 1
    Save 722.42 €
    36% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Supplied air regulator heavy industry kit provides continuous compressed air flow with adjustable control for durable, comfortable respiratory protection.
  • Delivers a continuous flow of air from a compressed air source to ensure a steady, consistent supply.
  • Compatible with a wide range of 3M headtops, including hoods and helmets, offering versatility for multiple applications.
  • Features adjustable airflow control, allowing users to customize airflow according to their individual needs and preferences.
  • Designed to protect against airborne contaminants for enhanced safety and performance.
  • Ideal for use in industries such as manufacturing, construction, and chemical processing.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Product Type Supplied Air Regulator