Learn More
3M™ Versaflo™ Supplied Air Regulator Paint Hood Kit, V-500E-PHK
Product Code. 31605230 Shop All 3M Products
Change view
Click to view available options
Product Type:
Supplied Air Regulator
Unit Size:
Each
Product Code. Product Type Unit Size
31605230 Supplied Air Regulator Each
1 options
Product Code. 31605230 Supplier 3M™ Supplier No. 7100342201
  • ONLINE EXCLUSIVE PRICE
    1430.00 €
    914.98 € / Pack of 1
    Save 515.02 €
    36% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
V-500E Series supplied air regulator assembly provides reliable respiratory protection with adjustable airflow and continuous compressed air for comfort and durability in tough environments.
  • 3M™ Versaflo™ Supplied Air Respirator System V-500E Series provides dependable respiratory protection and comfort for workers in diverse industrial settings.
  • Delivers a continuous flow of air from a compressed air source, ensuring a steady and reliable air supply.
  • Compatible with a wide range of 3M headtops, including hoods and helmets, for versatile application.
  • Features adjustable airflow control, allowing users to customize airflow based on individual needs and preferences.
  • Protects against airborne contaminants to enhance safety and performance.
  • Ideal for use in industries such as manufacturing, construction, and chemical processing.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Product Type Supplied Air Regulator