Learn More
3M™ Versaflo™ TR-600 Series Filter Covers
For use with Versaflo™ TR-600 Series PAPR systems.
Brand: 3M™ 7100021252
13.40 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 7.00000kg
Description
- Filter cover helps protect cartridge from physical damage
- Easy to use cover enables use of prefilters and/or spark arrestors
- Allows PAPR to be worn through decontamination showers or rain
- Also used to hold TR-6600 Prefilter or TR-662 Spark Arrestor in place
- Color: Gray
- 3M Catalog# TR-6500FC , 3M Catalog# TR-6700FC
Applications:
Recommended Industries: Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifications
NIOSH | |
Filter Cover | |
TR-6500 and TR-600 Respirators, TR-6600 Prefilter, and TR-662 Spark Arrestor | |
1/Cs. |
Gray | |
Healthcare, Pharmaceutical | |
TR-6500FC |