missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ TR-Series PAPR Accessory, Backpack

Product Code. 11743916
Change view
Click to view available options
:
Backpack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11743916 Backpack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11743916 Supplier 3M™ Supplier No. 7100005724

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allows for the TR-600 series PAPR to be work on the wearer's back rather than on a belt at the waist.

  • Provides for comfortable and convenient wear
  • Allows for additional equipment to be worn around the waist
  • Multi-layer construction for durability 
  • NIOSH approved
  • 3M Catalog# BPK-01

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Backpack
For Use With (Application) Animal processing, cosmetics, dry foods, general manufacturing, healthcare, heavy industrial, laboratories, manufacturing, nuclear energy, pesticide application, pharmaceutical, research and production, recycling
For Use With (Equipment) Versaflo TR-Series Powered Air Purifying Respirator
Model BKP-01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.