missing translation for 'onlineSavingsMsg'
Learn More

(3R)-Methyl 4-((1bS,3R)-7-(4-Chlorophenyl)-7-hydroxy-1b-isopropyl-7a-phenyl-1a-(phenylcarbamoyl)hexahydro-1aH-oxireno[2',3':3,4]pyrrolo[2,1-b][1,3]oxazin-3-yl)-3-hydroxybutanoate, TRC

Product Code. 30475661
Change view
Click to view available options
Quantity:
10 mg
250 mg
500 mg
Unit Size:
10mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475661 250 mg 250mg
30482779 10 mg 10mg
30484218 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475661 Supplier Toronto Research Chemicals Supplier No. TRCM340598250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R)-Methyl 4-((1bS,3R)-7-(4-Chlorophenyl)-7-hydroxy-1b-isopropyl-7a-phenyl-1a-(phenylcarbamoyl)hexahydro-1aH-oxireno[2',3':3,4]pyrrolo[2,1-b][1,3]oxazin-3-yl)-3-hydroxybutanoate
Analyte or Component Names (3R)-Methyl 4-((1bS,3R)-7-(4-Chlorophenyl)-7-hydroxy-1b-isopropyl-7a-phenyl-1a-(phenylcarbamoyl)hexahydro-1aH-oxireno[2',3':3,4]pyrrolo[2,1-b][1,3]oxazin-3-yl)-3-hydroxybutanoate
Shipping Condition Room Temperature
Molecular Formula C34 H37 Cl N2 O7
InChI Formula InChI=1S/C34H37ClN2O7/c1-22(2)34-32(30(40)36-26-12-8-5-9-13-26)31(44-32,23-10-6-4-7-11-23)33(41,24-14-16-25(35)17-15-24)37(34)19-18-28(43-34)20-27(38)21-29(39)42-3/h4-17,22,27-28,38,41H,18-21H2,1-3H3,(H,36,40)/t27-,28-,31?,32?,33?,34+/m1/s1
Solution Type Neat
SMILES COC(=O)C[C@H](O)C[C@H]1CCN2C(O)(c3ccc(Cl)cc3)C4(OC4(C(=O)Nc5ccccc5)[C@@]2(O1)C(C)C)c6ccccc6
IUPAC Name methyl (3R)-4-[(1S,9R)-5-(4-chlorophenyl)-5-hydroxy-4-phenyl-2-(phenylcarbamoyl)-1-propan-2-yl-3,10-dioxa-6-azatricyclo[4.4.0.02,4]decan-9-yl]-3-hydroxybutanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 621.12
Quantity 250 mg
Formula Weight 620.229
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.