missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Thiazol-2-yl)benzaldehyde, 95%, Thermo Scientific™

Product Code. 10576314
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10576314 250 mg 250mg
10439953 1 g 1g
10193214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10576314 Supplier Thermo Scientific Maybridge Supplier No. CC40204CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 198904-53-9
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.23
MDL Number MFCD06797780
InChI Key LQLBILPEELCFQI-UHFFFAOYSA-N
PubChem CID 10535655
IUPAC Name 4-(1,3-thiazol-2-yl)benzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=NC=CS1

Specifications

Assay Percent Range 95%
Quantity 250 mg
Synonym 4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde
Formula Weight 189.24
Percent Purity 95%
Chemical Name or Material 4-(1, 3-Thiazol-2-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.