missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Cyanoethyl)-1,2,3,4-tetrahydronaphthalene-1-carbonitrile, TRC

Product Code. 17964125
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964125 5 mg 5mg
17951415 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17964125 Supplier Toronto Research Chemicals Supplier No. TRCC9795455MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57964-39-3
Molecular Formula C14 H14 N2
Molecular Weight (g/mol) 210.27
Synonym 4-(1-Cyanoethyl)-1,2,3,4-tetrahydronaphthalene-1-carbonitrile, 4-Cyano-1,2,3,4-tetrahydro-α-methyl-1-naphthaleneacetonitrile, 1-Cyano-4-(1-cyanoethyl)-1,2,3,4-tetrahydronaphthalene
IUPAC Name 4-(1-cyanoethyl)tetralin-1-carbonitrile
SMILES CC(C#N)C1CCC(C#N)c2ccccc12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H14N2/c1-10(8-15)12-7-6-11(9-16)13-4-2-3-5-14(12)13/h2-5,10-12H,6-7H2,1H3
Formula Weight 210.1157
Chemical Name or Material 4-(1-Cyanoethyl)tetralin-1-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.