missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde 97+%, Thermo Scientific™

Product Code. 10799953
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10799953 250 mg Amber Glass Bottle 250mg
10127792 1 g Amber Glass Bottle 1g
10683635 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10799953 Supplier Thermo Scientific Maybridge Supplier No. CC45704CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 859850-94-5
Molecular Formula C10H9N3O
Molecular Weight (g/mol) 187.202
InChI Key XEPIORKIFKJATD-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazolylmethyl benzaldehyde, 4-1h-1,2,4-triazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-1,2,4-triazol-1-ylmethyl
PubChem CID 7164655
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC=C1CN2C=NC=N2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.