missing translation for 'onlineSavingsMsg'
Learn More

4-(1h-Imidazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10366664
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10366664 250 mg 250mg
10327763 1 g 1g
10451324 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366664 Supplier Thermo Scientific Maybridge Supplier No. CC43304CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 102432-03-1
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
InChI Key PYVCQHSLEIAQET-UHFFFAOYSA-N
PubChem CID 11126983
IUPAC Name 4-(imidazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC=C1CN2C=CN=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-1h-imidazol-1-ylmethyl benzaldehyde, 4-imidazol-1-ylmethyl benzaldehyde, 4-1h-imidazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-imidazol-1-ylmethyl, acmc-20e4oe, 4-imidazolylmethyl benzaldehyde, benzaldehyde, 4-1h-imidazol-1-ylmethyl
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.