missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate, 97%, Thermo Scientific™

Product Code. 10235564
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10235564 250 mg Amber Glass Bottle 250mg
10506334 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235564 Supplier Thermo Scientific Maybridge Supplier No. CC43396CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-84-9
Molecular Formula C14H17N3O4
Molecular Weight (g/mol) 291.307
MDL Number MFCD09064952
InChI Key AIQAEMDKVTYASZ-UHFFFAOYSA-N
Synonym oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
PubChem CID 24229491
IUPAC Name 1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid
SMILES CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 345.34
Percent Purity 95%
Chemical Name or Material 4-(1H-imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.