missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)benzylamine hydrochloride , Tech., Thermo Scientific™

Product Code. 10386704
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10386704 250 mg Amber Glass Bottle 250mg
10533455 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386704 Supplier Thermo Scientific Maybridge Supplier No. CC56063CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 904696-62-4
Molecular Formula C11H14ClN3
Molecular Weight (g/mol) 223.70
MDL Number MFCD07690519
InChI Key QUVMWCVEYSYIOW-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl methyl phenyl methanamine hydrochloride, 4-1h-pyrazol-1-ylmethyl benzylamine hydrochloride, 4-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 1-4-1h-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 1-4-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 4-pyrazolylmethyl phenyl methylamine, chloride, 4-1h-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 4-1h-pyrazol-1-yl methyl phenyl methanaminehydrochloride, 1-4-1h-pyrazol-1-yl methyl phenyl methanamine-hydrogen chloride 1/1
PubChem CID 16269819
IUPAC Name [4-(pyrazol-1-ylmethyl)phenyl]methanamine;hydrochloride
SMILES Cl.NCC1=CC=C(CN2C=CC=N2)C=C1

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 223.71
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(1H-pyrazol-1-ylmethyl)benzylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.