missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10534984
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10534984 5 g 5g
10131312 250 mg 250mg
10652253 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10534984 Supplier Thermo Scientific Maybridge Supplier No. CC25509DE
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 143426-51-1
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD03659708
InChI Key LQQQPLUFBVYLRE-UHFFFAOYSA-N
Synonym 4-1h-pyrrol-1-yl phenyl methanol, 1-4-hydroxymethylphenyl pyrrole, 4-1h-pyrrol-1-yl benzyl alcohol, 4-pyrrol-1-yl phenyl methanol, benzenemethanol,4-1h-pyrrol-1-yl, 4-pyrrolylphenyl methan-1-ol, 4-1h-pyrrol-1-yl benzylalcohol, pubchem11303, 4-pyrrol-1-ylphenyl methanol
PubChem CID 599478
IUPAC Name (4-pyrrol-1-ylphenyl)methanol
SMILES C1=CN(C=C1)C2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [4-(1H-Pyrrol-1-yl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.