missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine, 95%, Thermo Scientific™

Product Code. 11330667
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11330667 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11330667 Supplier Thermo Scientific Maybridge Supplier No. CC48639CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 876316-33-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C17H26BNO3
MDL Number MFCD07781208
Quantity 250 mg
Synonym 4-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 2-4-morpholinomethyl phenylboronic acid pinacol ester, 2-morpholinomethyl phenylboronic acid pinacol ester, 2-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 2-4-morpholinylmethyl benzeneboronic acid pinacol ester, 4-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, amtb374, 2-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester
InChI Key WCNXVDLZTGODQW-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1CN1CCOCC1
IUPAC Name 4-[[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
Molecular Weight (g/mol) 303.21
PubChem CID 16414146
Formula Weight 303.21
Percent Purity 95%
Chemical Name or Material 4-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.