missing translation for 'onlineSavingsMsg'
Learn More

4'-(2,5-Dimethoxyphenylsulfamoyl)acetanilide, 97%, Thermo Scientific™

Product Code. 15477138
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477138 1 g 1g
15425936 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477138 Supplier Thermo Scientific Alfa Aesar Supplier No. H60663.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool dry ventilated place. Ensure proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211742-82-4
Molecular Formula C16H18N2O5S
Molecular Weight (g/mol) 350.389
MDL Number MFCD01134327
InChI Key VVYBOPRFGIFZSH-UHFFFAOYSA-N
Synonym n-4-2,5-dimethoxyphenyl sulfamoyl phenyl acetamide, n-4-2,5-dimethoxy-phenylsulfamoyl-phenyl-acetamide, n-4-2,5-dimethoxyanilino sulfonyl phenyl acetamide, n-4-2,5-dimethoxyphenyl amino sulfonyl phenyl acetamide, 4'-2,5-dimethoxyphenylsulfamoyl acetanilide, 4/'-2,5-dimethoxyphenylsulfamoyl acetanilide
PubChem CID 1010065
IUPAC Name N-[4-[(2,5-dimethoxyphenyl)sulfamoyl]phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC(=C2)OC)OC

Specifications

Quantity 1 g
Formula Weight 350.39
Percent Purity 97%
Chemical Name or Material 4'-(2,5-Dimethoxyphenylsulfamoyl)acetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.