missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Carboxyethyl)benzeneboronic acid, 97%

Product Code. 11328456
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328456 5 g 5g
11318456 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328456 Supplier Thermo Scientific Alfa Aesar Supplier No. L17485.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 166316-48-9 | C9H11BO4 | 193.993 g/mol

4-(2-Carboxyethyl)benzeneboronic acid is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Carboxyethyl)benzeneboronic acid is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166316-48-9
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.993
MDL Number MFCD01318119
InChI Key VPSARXNVXCRDIV-UHFFFAOYSA-N
Synonym 3-4-boronophenyl propanoic acid, 4-2-carboxyethyl phenylboronic acid, 4-2-carboxyethyl benzeneboronic acid, 3-4-boronophenyl propionic acid, 3-4-dihydroxyboranyl phenyl propanoic acid, acmc-209dty, benzenepropanoic acid,4-borono, 3-4-boronophenyl-propanoic acid, 4-borono-benzenepropanoic acid, 4-2-carboxyethyl phenyl boronic acid
PubChem CID 3863484
IUPAC Name 3-(4-boronophenyl)propanoic acid
SMILES B(C1=CC=C(C=C1)CCC(=O)O)(O)O

Specifications

Melting Point 146°C to 148°C
Quantity 5 g
Beilstein 9198524
Solubility Information Slightly soluble in water.
Formula Weight 193.99
Percent Purity 97%
Chemical Name or Material 4-(2-Carboxyethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.