missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloro-6-nitrophenoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10496452
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10496452 1 g 1g
10772843 10 g 10g
10371765 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10496452 Supplier Thermo Scientific Maybridge Supplier No. BTB02778DA
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-00-9
Molecular Formula C12H7Cl2NO5S
Molecular Weight (g/mol) 348.15
MDL Number MFCD00052679
InChI Key CRTUVOFOPIFTQS-UHFFFAOYSA-N
Synonym 4-2-chloro-6-nitrophenoxy benzene-1-sulfonyl chloride, 4-2-chloro-6-nitrophenoxy benzenesulfonyl chloride, 4-6-nitro-2-chlorophenoxy-benzenesulfonyl chloride, 4-2-chloro-6-nitrophenoxy benzene-1-sulphonyl chloride
PubChem CID 2774276
IUPAC Name 4-(2-chloro-6-nitrophenoxy)benzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)OC2=CC=C(C=C2)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 348.16
Percent Purity 97%
Chemical Name or Material 4-(2-Chloro-6-nitrophenoxy)benzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.