missing translation for 'onlineSavingsMsg'
Learn More

[[4-[2-(Cyclopropylmethoxy)ethyl]phenoxy]methyl]oxirane, TRC

Product Code. 30353769
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353769 25 mg 25mg
30362069 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30353769 Supplier Toronto Research Chemicals Supplier No. TRCC98867025MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63659-17-6
Molecular Formula C15 H20 O3
Molecular Weight (g/mol) 248.32
Synonym Betaxolol Hydrochloride Imp. C (EP), Betaxolol Imp. C (EP), 2-[[4-[2-(Cyclopropylmethoxy)ethyl]phenoxy]methyl]oxirane, Betaxolol Hydrochloride Impurity C, Betaxolol Impurity C
IUPAC Name 2-[[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]methyl]oxirane
SMILES C(Cc1ccc(OCC2CO2)cc1)OCC3CC3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H20O3/c1-2-13(1)9-16-8-7-12-3-5-14(6-4-12)17-10-15-11-18-15/h3-6,13,15H,1-2,7-11H2
Formula Weight 248.1412
Chemical Name or Material 2-[[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]methyl]oxirane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.