missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Dimethylamino)ethoxy]benzylamine, TRC

Product Code. 30008469
Change view
Click to view available options
Quantity:
250 mg
1 g
100 g
Unit Size:
100g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008469 1 g 1g
30009832 100 g 100g
30005080 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008469 Supplier Toronto Research Chemicals Supplier No. TRCD4711651G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20059-73-8
Molecular Formula C11 H18 N2 O
Molecular Weight (g/mol) 194.27
Synonym Benzenemethanamine, 4-[2-(dimethylamino)ethoxy]- (9CI, ACI), 4-[2-(Dimethylamino)ethoxy]benzenemethanamine (ACI), Benzylamine, p-[2-(dimethylamino)ethoxy]- (6CI, 7CI, 8CI), 2-[4-(Aminomethyl)phenoxy]-N,N-dimethylethan-1-amine, 2-[4-(Aminomethyl)phenoxy]-N,N-dimethylethanamine, 4-[2-(Dimethylamino)ethoxy]benzylamine, [2-(4-Aminomethyl-phenoxy)-ethyl]-dimethyl-amine, [2-[(4-Aminomethylphenyl)oxy]ethyl]dimethylamine, [4-[2-(Dimethylamino)ethoxy]phenyl]methanamine, NSC 37857
IUPAC Name 2-[4-(aminomethyl)phenoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOc1ccc(CN)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H18N2O/c1-13(2)7-8-14-11-5-3-10(9-12)4-6-11/h3-6H,7-9,12H2,1-2H3
Formula Weight 194.1419
Chemical Name or Material 4-[2-(Dimethylamino)ethoxy]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.