missing translation for 'onlineSavingsMsg'
Learn More

4-(2-furyl)benzaldehyde, Thermo Scientific™

Product Code. 10432453
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10432453 1 g 1g
10085952 250 mg 250mg
10372055 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10432453 Supplier Thermo Scientific Maybridge Supplier No. CC34404DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 60456-77-1
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
InChI Key WBUXKMOCVYRVES-UHFFFAOYSA-N
Synonym 4-2-furyl benzaldehyde, 4-furan-2-yl benzaldehyde, 4-furan-2-yl-benzaldehyde, benzaldehyde,4-2-furanyl, 2-4-formylphenyl furan, 4-2-furanyl benzaldehyde
PubChem CID 2772295
IUPAC Name 4-(furan-2-yl)benzaldehyde
SMILES C1=COC(=C1)C2=CC=C(C=C2)C=O

Specifications

Quantity 1 g
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material 4-(2-furyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.