missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)phenol, 97%, Thermo Scientific™

Product Code. 10302055
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10302055 5 g 5g
10276004 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10302055 Supplier Thermo Scientific Maybridge Supplier No. SB01196DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 501-94-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002902
InChI Key YCCILVSKPBXVIP-UHFFFAOYSA-N
Synonym tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
PubChem CID 10393
ChEBI CHEBI:1879
IUPAC Name 4-(2-hydroxyethyl)phenol
SMILES C1=CC(=CC=C1CCO)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula HOC6H4CH2CH2OH
Quantity 5 g
Merck Index 15, 10022
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 4-(2-Hydroxyethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.