missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methylpropyl)benzeneethanol, TRC

Product Code. 30478078
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478078 250 mg 250mg
30474094 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478078 Supplier Toronto Research Chemicals Supplier No. TRCM326900250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36039-35-7
Molecular Formula C12 H18 O
Molecular Weight (g/mol) 178.27
Synonym 2-[4-(2-Methylpropyl)phenyl]ethanol, Ibuprofen Imp. Q (EP)
IUPAC Name 2-[4-(2-methylpropyl)phenyl]ethanol
SMILES CC(C)Cc1ccc(CCO)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H18O/c1-10(2)9-12-5-3-11(4-6-12)7-8-13/h3-6,10,13H,7-9H2,1-2H3
Formula Weight 178.1358
Chemical Name or Material 2-[4-(2-Methylpropyl)phenyl]ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.