missing translation for 'onlineSavingsMsg'
Learn More

[4-(2-Morpholinoethoxy)phenyl]methylamine, 95%, Thermo Scientific™

Product Code. 10138182
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10138182 250 mg Amber Glass Bottle 250mg
10452643 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10138182 Supplier Thermo Scientific Maybridge Supplier No. CC42113CB
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 140836-69-7
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.315
MDL Number MFCD07772849
InChI Key KMSJKJIIAVTIKY-UHFFFAOYSA-N
Synonym 4-2-morpholinoethoxy phenyl methylamine, 4-2-morpholinoethoxy phenyl methanamine, 4-2-morpholinoethoxy benzylamine, benzenemethanamine, 4-2-4-morpholinyl ethoxy, 4-2-morpholin-4-yl-ethoxy benzylamine, 4-2-morpholin-4-yl ethoxy phenyl methanamine, 1-4-2-morpholin-4-yl ethoxy phenyl methanamine, acmc-1bwvj, 4-2-morpholin-4-ylethoxy phenyl methanamine
PubChem CID 7164631
IUPAC Name [4-(2-morpholin-4-ylethoxy)phenyl]methanamine
SMILES C1COCCN1CCOC2=CC=C(C=C2)CN

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 236.31
Percent Purity 95%
Chemical Name or Material [4-(2-Morpholinoethoxy)phenyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.