missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]benzylamine, 97%, Thermo Scientific™

Product Code. 10773974
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10773974 1 g Amber Glass Bottle 1g
10597844 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10773974 Supplier Thermo Scientific Maybridge Supplier No. CC59513DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 91637-76-2
Molecular Formula C12H20N2O
Molecular Weight (g/mol) 208.305
MDL Number MFCD07786701
InChI Key UGICLWAMZSRUPM-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy benzylamine, 4-3-dimethylamino propoxy phenyl methanamine, 3-4-aminomethyl phenoxy-n,n-dimethylpropan-1-amine, 3-4-aminomethyl phenoxy propyl dimethylamine, benzenemethanamine, 4-3-dimethylamino propoxy, benzenemethanamine,4-3-dimethylamino propoxy, 3-4-aminomethyl phenoxy-n,n-dimethylprop an-1-amine, 1-4-3-dimethylamino propoxy phenyl methanamine
PubChem CID 21465384
IUPAC Name 3-[4-(aminomethyl)phenoxy]-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCOC1=CC=C(C=C1)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.3
Percent Purity 97%
Chemical Name or Material 4-[3-(Dimethylamino)propoxy]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.