missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine, 95%, Thermo Scientific™

Product Code. p-7033717
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459560 250 mg 250mg
11469560 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459560 Supplier Thermo Scientific Alfa Aesar Supplier No. H50874.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 912761-48-9
Molecular Formula C9H15N3O
Molecular Weight (g/mol) 181.239
MDL Number MFCD08061039
InChI Key PTQDWAPTOMWGGM-UHFFFAOYSA-N
PubChem CID 24208846
IUPAC Name 3-ethyl-5-piperidin-4-yl-1,2,4-oxadiazole
SMILES CCC1=NOC(=N1)C2CCNCC2

Specifications

Quantity 250 mg
Synonym 4-3-ethyl-1,2,4-oxadiazol-5-yl piperidine, 3-ethyl-5-4-piperidyl-1,2,4-oxadiazole, 3-ethyl-5-piperidin-4-yl-1,2,4-oxadiazole, piperidine, 4-3-ethyl-1,2,4-oxadiazol-5-yl, pubchem18474, 4-3-ethyl-1,2,4 oxadiazol-5-yl-piperidine, piperidine,4-3-ethyl-1,2,4-oxadiazol-5-yl, 4-3-ethyl-1,2,4 oxadiazol-5-yl-piperidine hydrochloride
Solubility Information Slightly soluble in water.
Formula Weight 181.24
Percent Purity 95%
Chemical Name or Material 4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.