missing translation for 'onlineSavingsMsg'
Learn More

4,4'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis-benzeneacetamide (Atenolol Impurity E), TRC

Product Code. 30137951
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137951 100 mg 100mg
30010205 25 mg 25mg
17924055 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137951 Supplier Toronto Research Chemicals Supplier No. TRCH948995100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141650-31-9
Molecular Formula C19 H22 N2 O5
Molecular Weight (g/mol) 358.39
Synonym 2,2'-[(2-Hydroxypropane-1,3-diyl)bis(oxy-4,1-phenylene)]diacetamide, Atenolol Imp. E (EP), Benzeneacetamide, 4,4'-[(2-hydroxy-1,3-propanediyl)bis(oxy)]bis-, 4,4'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[benzeneacetamide]
IUPAC Name 2-[4-[3-[4-(2-amino-2-oxoethyl)phenoxy]-2-hydroxypropoxy]phenyl]acetamide
SMILES NC(=O)Cc1ccc(OCC(O)COc2ccc(CC(=O)N)cc2)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H22N2O5/c20-18(23)9-13-1-5-16(6-2-13)25-11-15(22)12-26-17-7-3-14(4-8-17)10-19(21)24/h1-8,15,22H,9-12H2,(H2,20,23)(H2,21,24)
Formula Weight 358.1529
Percent Purity >95%
Chemical Name or Material 2,2'-[(2-Hydroxypropane-1,3-diyl)bis(oxy-4,1-phenylene)]diacetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.