missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Trihydroxytriphenylmethane, 97%, Thermo Scientific™

Product Code. 15294794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15294794 100 g 100g
15284794 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15294794 Supplier Thermo Scientific Acros Supplier No. 450101000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 603-44-1
Molecular Formula C19H16O3
Molecular Weight (g/mol) 292.33
MDL Number MFCD00191589
InChI Key WFCQTAXSWSWIHS-UHFFFAOYSA-N
Synonym 4,4',4-methanetriyltriphenol, leucoaurin, 4,4',4-trihydroxytriphenylmethane, leucoaurine, leucaurin, phenol, methylidynetris, 4-bis 4-hydroxyphenyl methyl phenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, phenol, 4,4',4-methylidynetris
PubChem CID 69047
IUPAC Name 4-[bis(4-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 244.0°C to 246.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 292.33
Percent Purity 97%
Chemical Name or Material 4, 4', 4''-Trihydroxytriphenylmethane
compliance-icons
Product Identifier
  • 4,4
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.