missing translation for 'onlineSavingsMsg'
Learn More

4-(4'-Aminobenzyl)thiomorpholine 1,1-dioxide, ≥90%, Thermo Scientific™

Product Code. 10384482
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10384482 250 mg 250mg
10292932 1 g 1g
10108504 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10384482 Supplier Thermo Scientific Maybridge Supplier No. MO00124CB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 263339-24-8
Molecular Formula C11H16N2O2S
Molecular Weight (g/mol) 240.32
MDL Number MFCD03407300
InChI Key TZMCILRCGMFRJC-UHFFFAOYSA-N
Synonym 4-4'-aminobenzyl thiomorpholine 1,1-dioxide, 4-1,1-dioxo-1,4-thiazinan-4-yl methyl aniline, 4-1,1-dioxo-4-thiomorpholinyl methyl phenyl amine, 4-4-aminophenyl methyl-1??-thiomorpholine-1,1-dione, 4-aminobenzyl thiomorpholine 1,1-dioxide, 4-4-aminobenzyl thiomorpholine 1,1-dioxide, 4-4-aminobenzyl-thiomorpholine-1,1-dioxide, 4-4-amino-benzyl-thiomorpholine 1,1-dioxide, 4-1,1-dioxo-thiomorpholin-4-yl-methyl-aniline
PubChem CID 2779682
IUPAC Name 4-[(1,1-dioxo-1,4-thiazinan-4-yl)methyl]aniline
SMILES NC1=CC=C(CN2CCS(=O)(=O)CC2)C=C1

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 240.33
Percent Purity 90%
Chemical Name or Material 4-(4'-Aminobenzyl)thiomorpholine 1, 1-dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.