missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine, 95%, Thermo Scientific™

Product Code. p-7029448
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15407635 250 mg 250mg
11491470 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407635 Supplier Thermo Scientific Alfa Aesar Supplier No. H26911.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885952-23-8
Molecular Formula C15H16BrN3O2
Molecular Weight (g/mol) 350.216
MDL Number MFCD06798240
InChI Key DMIIRAXOFYSJQT-UHFFFAOYSA-N
Synonym 4-4-benzyloxy-5-bromopyrimidin-2-yl morpholine, 4-4-benzyloxy-5-bromo-2-pyrimidinyl morpholine, 4-benzyloxy-5-bromo-2-morpholin-4-yl-pyrimidine, morpholine,4-5-bromo-4-phenylmethoxy-2-pyrimidinyl
PubChem CID 42552971
IUPAC Name 4-(5-bromo-4-phenylmethoxypyrimidin-2-yl)morpholine
SMILES C1COCCN1C2=NC=C(C(=N2)OCC3=CC=CC=C3)Br

Specifications

Melting Point 114°C to 116°C
Quantity 250 mg
Formula Weight 350.21
Percent Purity 95%
Chemical Name or Material 4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.