missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenol, 97%

Product Code. 10452845 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
10g
250g
25g
Product Code. Quantity Packaging unitSize
10452845 10 g Glass bottle 10g
10782021 25 g Plastic bottle 25g
10614852 100 g Plastic bottle 100g
10217163 250 g Plastic bottle 250g
4 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 10452845

Brand: Thermo Scientific Acros 173670100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-88-6 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-88-6
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002348
InChI Key VCCBEIPGXKNHFW-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybiphenyl, 4,4'-biphenol, 4,4'-biphenyldiol, biphenyl-4,4'-diol, 1,1'-biphenyl-4,4'-diol, p,p'-biphenol, p,p'-diphenol, antioxidant dod, 4,4'-dioxydiphenyl, p-dihydroxydiphenyl
PubChem CID 7112
ChEBI CHEBI:34367
IUPAC Name 4-(4-hydroxyphenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 278°C to 283°C
Color Beige to White or Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H10O2
Linear Formula HOC6H4C6H4OH
Quantity 10 g
Beilstein 06, 991
Solubility Information Solubility in water: sparingly soluble
Formula Weight 186.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4, 4'-Biphenol
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Biphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.