missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenyldicarboxylic acid, 98%

Product Code. 10463365 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10463365 5 g Glass bottle 5g
10772264 1 g Glass bottle 1g
10360864 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10463365 Supplier Thermo Scientific Acros Supplier No. 387300050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 787-70-2 | C14H10O4 | 242.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 787-70-2
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00002554
InChI Key NEQFBGHQPUXOFH-UHFFFAOYSA-N
Synonym biphenyl-4,4'-dicarboxylic acid, 4,4'-biphenyldicarboxylic acid, 1,1'-biphenyl-4,4'-dicarboxylic acid, 4-4-carboxyphenyl benzoic acid, 4,4'-bibenzoic acid, 4,4'-diphenic acid, 4,4'-dicarboxybiphenyl, p,p'-diphenic acid, usaf do-69, bibenzoic acid
PubChem CID 13084
IUPAC Name 4-(4-carboxyphenyl)benzoic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 300.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H4COOH)2
Quantity 5 g
Formula Weight 242.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Biphenyldicarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Biphenyldicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.