missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(2-sulfostyryl)biphenyl Disodium, TRC

Product Code. 17994825
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994825 100 g 100g
17943285 10 g 10g
17993735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994825 Supplier Toronto Research Chemicals Supplier No. TRCB535200100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27344-41-8
Molecular Formula C28 H20 O6 S2 . 2 Na
Molecular Weight (g/mol) 562.56
Synonym 4,4'-Bis(o-sulfostyryl)biphenyl Disodium, Benzenesulfonic acid, 2,2'-(4,4'-biphenylylenedivinylene)di-, disodium salt (8CI), Benzenesulfonic acid, 2,2'-([1,1'-biphenyl]-4,4'-diyldi-2,1-ethenediyl)bis-, disodium salt (9CI), 4,4'-Bis(2-sodiosulfostyryl)biphenyl, 4,4'-Bis(2-sulfostyryl)biphenyl disodium salt, 4,4'-Bis(o-sulfostyryl)biphenyl disodium salt, Benetex OB-M 1, Brightener BR 49, Brightener CBS-X, CBS-X, CF 351, Calcofluor CG, DSBP, Disodium 2,2'-(4,4'-biphenyldiyldi-2,1-ethenediyl)dibenzenesulfonate, Disodium 2,2'-([1,1'-biphenyl]-4,4'-diyldivinylene)bis(benzenesulfonate), Disodium 4,4'-bis(2-sulfostyryl)biphenyl, Disodium 4',4' '-bi[stilbene-2,2' ''-disulfonate], FWA-CBS-X, Keyfluor CBS-X, Keyfluor White CBS-X, OB-M 1, Optical Brightener CBS-X, Stilbene 3, Stilbene 420, Stilbene S-420, Tinopal CBS, Tinopal CBS-X, Tinopal CBS-X 44, Tinopal CBX, Tinopal NFW, Tinopal NFW Liq, Tinopal SK, Uvitex CBS-X, Uvitex NFW, Benzenesulfonic acid, 2,2'-([1,1'-biphenyl]-4,4'-diyldi-2,1-ethenediyl)bis-, sodium salt (1:2)
IUPAC Name disodium;2-[(E)-2-[4-[4-[(E)-2-(2-sulfonatophenyl)ethenyl]phenyl]phenyl]ethenyl]benzenesulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)c1ccccc1\C=C\c2ccc(cc2)c3ccc(\C=C\c4ccccc4S(=O)(=O)[O-])cc3

Specifications

Quantity 100 g
InChI Formula InChI=1S/C28H22O6S2.2Na/c29-35(30,31)27-7-3-1-5-25(27)19-13-21-9-15-23(16-10-21)24-17-11-22(12-18-24)14-20-26-6-2-4-8-28(26)36(32,33)34;;/h1-20H,(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2/b19-13+,20-14+;;
Formula Weight 562.0497
Chemical Name or Material 4,4'-Bis(2-sulfostyryl)biphenyl Disodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.