missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dimethylamino)benzophenone, 98%

Product Code. 10225500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225500 5 g Glass bottle 5g
10317730 100 g Glass bottle 100g
10615032 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10225500 Supplier Thermo Scientific Acros Supplier No. 164490050
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 90-94-8 | C17H20N2O | 268.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-94-8
Molecular Formula C17H20N2O
Molecular Weight (g/mol) 268.36
MDL Number MFCD00008312
InChI Key VVBLNCFGVYUYGU-UHFFFAOYSA-N
Synonym michler's ketone, 4,4'-bis dimethylamino benzophenone, michler ketone, bis 4-dimethylamino phenyl methanone, p,p'-michler's ketone, michlers ketone, methanone, bis 4-dimethylamino phenyl, tetramethyldiaminobenzophenone, p,p'-bis dimethylamino benzophenone
PubChem CID 7031
ChEBI CHEBI:82347
IUPAC Name bis[4-(dimethylamino)phenyl]methanone
SMILES CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 171°C to 176°C
Color Blue or Gray
Boiling Point >360°C
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula [(CH3)2NC6H4]2CO
Quantity 5 g
Beilstein 14, 89
Fieser 02,285
Merck Index 15, 6526
Solubility Information Solubility in water: 400mg/l (20°C). Other solubilities: soluble in alcohol,warm benzene and ether
Formula Weight 268.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Bis(dimethylamino)benzophenone, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4,4'-Bis(dimethylamino)benzophenone
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.