missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(Bromomethyl)phenyl]-1,2,3-thiadiazole, 97%, Thermo Scientific™

Product Code. 10732663
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10732663 1 g Amber Glass Bottle 1g
10582832 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732663 Supplier Thermo Scientific Maybridge Supplier No. CC16408DA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 163798-92-3
Molecular Formula C9H7BrN2S
Molecular Weight (g/mol) 255.13
MDL Number MFCD00052102
InChI Key DGHQOPZIGDRUIT-UHFFFAOYSA-N
Synonym 4-4-bromomethyl phenyl-1,2,3-thiadiazole, 1,2,3-thiadiazole,4-4-bromomethyl phenyl, 4-4-bromomethyl phenyl thiadiazole, 4-1,2,3-thiadiazol-4-yl benzyl bromide, 4-4-bromomethylphenyl-1,2,3-thiadiazole, 4-4-bromomethyl-phenyl-1,2,3 thiadiazole
PubChem CID 2776454
IUPAC Name 4-[4-(bromomethyl)phenyl]thiadiazole
SMILES BrCC1=CC=C(C=C1)C1=CSN=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 255.14
Percent Purity 97%
Chemical Name or Material 4-[4-(Bromomethyl)phenyl]-1, 2, 3-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.