missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)-1,2,3-thiadiazole, 97%, Thermo Scientific™

Product Code. 10772093
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772093 1 g Amber Glass Bottle 1g
10522842 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772093 Supplier Thermo Scientific Maybridge Supplier No. CC16410DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 40753-13-7
Molecular Formula C8H5BrN2S
Molecular Weight (g/mol) 241.106
MDL Number MFCD00084907
InChI Key HGWOTVRPRHVJQK-UHFFFAOYSA-N
PubChem CID 218520
IUPAC Name 4-(4-bromophenyl)thiadiazole
SMILES C1=CC(=CC=C1C2=CSN=N2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-4-bromophenyl-1,2,3-thiadiazole, 1,2,3-thiadiazole, 4-4-bromophenyl, 1,2,3-thiadiazole,4-4-bromophenyl, 4-4-bromophenyl thiadiazole
Formula Weight 241.11
Percent Purity 97%
Chemical Name or Material 4-(4-Bromophenyl)-1, 2, 3-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.